Assay Details
| Gene Symbol: gabrz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114742.2 | NP_001108214.2 | 6 - 7 | 731 | 76 | |
| GenBank mRNA | BC159112.1 | - | 6 - 7 | 731 | 76 | |
More Information
Set Membership: |
|

gabrz
Genomic Map