Assay Details
| Gene Symbol: hexb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114845.2 | NP_001108317.1 | 13 - 14 | 1651 | 70 | |
| XM_005171791.4 | XP_005171848.1 | 13 - 14 | 1668 | 70 | ||
| XM_009301447.3 | XP_009299722.1 | 14 - 15 | 1674 | 70 | ||
| XM_009301448.3 | XP_009299723.1 | 14 - 15 | 1669 | 70 | ||
More Information
Set Membership: |
|

hexb
Genomic Map