Assay Details
| Gene Symbol: f2r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114846.1 | NP_001108318.1 | 1 - 2 | 69 | 115 | |
| GenBank mRNA | GQ325252.1 | - | 1 - 2 | 69 | 115 | |
More Information
Set Membership: |
|

f2r
Genomic Map