Assay Details
| Gene Symbol: mmachc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114889.2 | NP_001108361.2 | 3 - 4 | 493 | 89 | |
| GenBank mRNA | BC154014.1 | - | 3 - 4 | 449 | 89 | |
| EH554822.1 | - | 3 - 4 | 532 | 89 | ||
More Information
Set Membership: |
|

mmachc
Genomic Map