Assay Details
| Gene Symbol: bmpr1bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114924.1 | NP_001108396.1 | 5 - 6 | 578 | 85 | |
| NM_001145996.1 | NP_001139468.1 | 6 - 7 | 731 | 85 | ||
| GenBank mRNA | BC154593.1 | - | 5 - 6 | 578 | 85 | |
| FJ655416.1 | - | 6 - 7 | 731 | 85 | ||
More Information
Set Membership: |
|

bmpr1bb
Genomic Map