Assay Details
Gene Symbol: igf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115050.1 | NP_001108522.1 | 2 - 3 | 250 | 99 | |
GenBank mRNA | EU272146.1 | - | 2 - 3 | 250 | 99 | |
HQ241070.1 | - | 2 - 3 | 250 | 99 | ||
HQ241071.1 | - | 2 - 3 | 378 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
