Assay Details
| Gene Symbol: tgfbr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115059.1 | NP_001108531.1 | 5 - 6 | 1040 | 92 | |
| GenBank mRNA | EF462418.1 | - | 5 - 6 | 1040 | 92 | |
More Information
Set Membership: |
|

tgfbr1b
Genomic Map