Assay Details
| Gene Symbol: tbr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115090.3 | NP_001108562.1 | 1 - 2 | 761 | 90 | |
| GenBank mRNA | BC168510.1 | - | 1 - 2 | 761 | 90 | |
More Information
Set Membership: |
|

tbr1b
Genomic Map