Assay Details
| Gene Symbol: tbr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115090.3 | NP_001108562.1 | 2 - 3 | 920 | 60 | |
| GenBank mRNA | AF287006.1 | - | 1 - 2 | 122 | 60 | |
| BC168510.1 | - | 2 - 3 | 920 | 60 | ||
More Information
Set Membership: |
|

tbr1b
Genomic Map