Assay Details
| Gene Symbol: tbr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115090.3 | NP_001108562.1 | 5 - 6 | 1269 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

tbr1b| Gene Symbol: tbr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115090.3 | NP_001108562.1 | 5 - 6 | 1269 | 64 | |
Set Membership: |
|