Assay Details
| Gene Symbol: gapdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115114.1 | NP_001108586.1 | 6 - 7 | 507 | 121 | |
| GenBank mRNA | BC083506.1 | - | 6 - 7 | 507 | 121 | |
| BC095386.1 | - | 6 - 7 | 507 | 121 | ||
| BC115131.1 | - | 6 - 7 | 511 | 121 | ||
More Information
Set Membership: |
|

gapdh
Genomic Map