Assay Details
| Gene Symbol: yap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001139480.1 | NP_001132952.1 | 6 - 7 | 1383 | 64 | |
| XM_005173662.4 | XP_005173719.1 | 5 - 6 | 1283 | 64 | ||
| GenBank mRNA | AI658050.1 | - | 3 - 4 | 459 | 64 | |
| BC129216.1 | - | 6 - 7 | 1352 | 64 | ||
More Information
Set Membership: |
|

yap1
Genomic Map