Assay Details
| Gene Symbol: dpp6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115122.1 | NP_001108594.1 | 8 - 9 | 937 | 69 | |
| XM_005163289.3 | XP_005163346.1 | 3 - 4 | 887 | 69 | ||
| XM_005163290.4 | XP_005163347.1 | 3 - 4 | 362 | 69 | ||
| XM_005163291.4 | XP_005163348.1 | 3 - 4 | 330 | 69 | ||
More Information
Set Membership: |
|

dpp6b
Genomic Map