Assay Details
| Gene Symbol: ewsr1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115138.2 | NP_001108610.1 | 11 - 12 | 1271 | 70 | |
| XM_021479109.1 | XP_021334784.1 | 11 - 12 | 1270 | 70 | ||
| GenBank mRNA | DT075886.1 | - | 1 - 2 | 50 | 70 | |
More Information
Set Membership: |
|

ewsr1a
Genomic Map