Assay Details
| Gene Symbol: ewsr1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115138.2 | NP_001108610.1 | 15 - 16 | 1873 | 69 | |
| XM_021479109.1 | XP_021334784.1 | 15 - 16 | 1872 | 69 | ||
| GenBank mRNA | BC068397.1 | - | 14 - 15 | 1842 | 69 | |
| DT075886.1 | - | 5 - 6 | 68 | |||
| FP131351.1 | - | 1 - 2 | 677 | 69 | ||
More Information
Set Membership: |
|

ewsr1a
Genomic Map