Assay Details
Gene Symbol: ppp4r2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115140.1 | NP_001108612.1 | 2 - 3 | 246 | 124 | |
GenBank mRNA | BC060912.1 | - | 2 - 3 | 184 | 124 | |
BC151885.1 | - | 2 - 3 | 205 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
