Assay Details
Gene Symbol: ppp4r2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115140.1 | NP_001108612.1 | 7 - 8 | 766 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
