Assay Details
Gene Symbol: magi2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001327853.1 | NP_001314782.1 | 16 - 17 | 3018 | 63 | |
XM_017355040.2 | XP_017210529.1 | 16 - 17 | 3059 | 63 | ||
XM_017355041.2 | XP_017210530.1 | 16 - 17 | 3056 | 63 | ||
XM_017355042.2 | XP_017210531.1 | 16 - 17 | 3059 | 63 | ||
XM_017355043.2 | XP_017210532.1 | 14 - 15 | 2853 | 63 | ||
XM_017355044.2 | XP_017210533.1 | 14 - 15 | 2851 | 63 | ||
XM_017355045.2 | XP_017210534.1 | 16 - 17 | 3065 | 63 | ||
XM_017355046.2 | XP_017210535.1 | 16 - 17 | 3064 | 63 | ||
XM_017355047.2 | XP_017210536.1 | 16 - 17 | 3065 | 63 | ||
XM_017355048.2 | XP_017210537.1 | 16 - 17 | 3065 | 63 | ||
XM_021475052.1 | XP_021330727.1 | 16 - 17 | 3061 | 63 | ||
GenBank mRNA | AI584740.1 | - | 2 - 3 | 373 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
