Assay Details
| Gene Symbol: ptbp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122654.1 | NP_001116126.1 | 2 - 3 | 161 | 90 | |
| GenBank mRNA | BC091854.1 | - | 2 - 3 | 123 | 90 | |
More Information
Set Membership: |
|

ptbp1b
Genomic Map