Assay Details
| Gene Symbol: smpd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122750.1 | NP_001116222.1 | 1 - 2 | 1400 | 60 | |
| XM_005163072.4 | XP_005163129.1 | 2 - 3 | 1838 | 60 | ||
| GenBank mRNA | AB361067.1 | - | 1 - 2 | 1400 | 60 | |
More Information
Set Membership: |
|

smpd3
Genomic Map