Assay Details
| Gene Symbol: matr3l1.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122784.2 | NP_001116256.2 | 20 - 21 | 3553 | 130 | |
| XM_005173122.4 | XP_005173179.1 | 20 - 21 | 3563 | 130 | ||
| XM_005173123.4 | XP_005173180.1 | 19 - 20 | 3515 | 130 | ||
| XM_009291035.3 | XP_009289310.1 | 19 - 20 | 2662 | 130 | ||
More Information
Set Membership: |
|

matr3l1.2
Genomic Map