Assay Details
| Gene Symbol: matr3l1.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122784.2 | NP_001116256.2 | 21 - 22 | 3656 | 92 | |
| GenBank mRNA | AI558298.1 | - | 1 - 2 | 337 | 92 | |
| AI558952.1 | - | 2 - 3 | 237 | 92 | ||
More Information
Set Membership: |
|

matr3l1.2
Genomic Map