Assay Details
| Gene Symbol: hmgb3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122836.1 | NP_001116308.1 | 2 - 3 | 271 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

hmgb3a| Gene Symbol: hmgb3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122836.1 | NP_001116308.1 | 2 - 3 | 271 | 81 | |
Set Membership: |
|