Assay Details
| Gene Symbol: derl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200903.2 | NP_957197.2 | 5 - 6 | 729 | 67 | |
| GenBank mRNA | BC064666.1 | - | 5 - 6 | 728 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

derl3| Gene Symbol: derl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200903.2 | NP_957197.2 | 5 - 6 | 729 | 67 | |
| GenBank mRNA | BC064666.1 | - | 5 - 6 | 728 | 67 | |
Set Membership: |
|