Assay Details
Gene Symbol: phkg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200962.1 | NP_957256.1 | 6 - 7 | 758 | 61 | |
XM_009306599.3 | XP_009304874.1 | 6 - 7 | 771 | 61 | ||
GenBank mRNA | BC047191.1 | - | 6 - 7 | 758 | 61 | |
BC075780.1 | - | 6 - 7 | 748 | 61 | ||
BC152636.1 | - | 6 - 7 | 718 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
