Assay Details
Gene Symbol: cpeb4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001351720.1 | NP_001338649.1 | 2 - 3 | 1245 | 74 | |
NM_200981.2 | NP_957275.2 | 2 - 3 | 1197 | 74 | ||
XM_005173113.4 | XP_005173170.2 | 2 - 3 | 1273 | 74 | ||
XM_009291021.3 | XP_009289296.1 | 2 - 3 | 1603 | 74 | ||
XM_017358999.2 | XP_017214488.1 | 2 - 3 | 1273 | 74 | ||
XM_017359000.2 | XP_017214489.1 | 2 - 3 | 1273 | 74 | ||
GenBank mRNA | BC055522.1 | - | 2 - 3 | 1197 | 74 | |
BC165168.1 | - | 1 - 2 | 940 | 74 | ||
EF033337.1 | - | 1 - 2 | 1392 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
