Assay Details
Gene Symbol: cpeb4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001351720.1 | NP_001338649.1 | 8 - 9 | 2015 | 75 | |
NM_200981.2 | NP_957275.2 | 8 - 9 | 1967 | 75 | ||
XM_005173113.4 | XP_005173170.2 | 9 - 10 | 2067 | 75 | ||
XM_009291021.3 | XP_009289296.1 | 9 - 10 | 2397 | 75 | ||
GenBank mRNA | BC055522.1 | - | 8 - 9 | 1967 | 75 | |
BC165168.1 | - | 7 - 8 | 1710 | 75 | ||
EF033337.1 | - | 7 - 8 | 2162 | 75 | ||
EH535381.1 | - | 1 - 2 | 467 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
