Assay Details
| Gene Symbol: acy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272080.1 | NP_001259009.1 | 12 - 13 | 1181 | 66 | |
| NM_200995.2 | NP_957289.2 | 12 - 13 | 1078 | 66 | ||
| XM_009296511.3 | XP_009294786.1 | 12 - 13 | 990 | 66 | ||
| GenBank mRNA | BC045399.1 | - | 12 - 13 | 1014 | 66 | |
| BC164807.1 | - | 11 - 12 | 988 | 66 | ||
More Information
Set Membership: |
|

acy1
Genomic Map