Assay Details
| Gene Symbol: mfap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201017.1 | NP_957311.1 | 3 - 4 | 532 | 97 | |
| GenBank mRNA | BC052118.1 | - | 3 - 4 | 532 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mfap1| Gene Symbol: mfap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201017.1 | NP_957311.1 | 3 - 4 | 532 | 97 | |
| GenBank mRNA | BC052118.1 | - | 3 - 4 | 532 | 97 | |
Set Membership: |
|