Assay Details
| Gene Symbol: prkacba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201023.1 | NP_957317.1 | 6 - 7 | 781 | 74 | |
| GenBank mRNA | BC053227.1 | - | 6 - 7 | 781 | 74 | |
| BC165279.1 | - | 6 - 7 | 715 | 74 | ||
More Information
Set Membership: |
|

prkacba
Genomic Map