Assay Details
| Gene Symbol: pgam2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201024.1 | NP_957318.1 | 2 - 3 | 692 | 103 | |
| GenBank mRNA | BC053127.1 | - | 2 - 3 | 692 | 103 | |
| BC171458.1 | - | 2 - 3 | 664 | 103 | ||
| BC171460.1 | - | 2 - 3 | 664 | 103 | ||
More Information
Set Membership: |
|

pgam2
Genomic Map