Assay Details
| Gene Symbol: amd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161601.1 | NP_001155073.1 | 1 - 2 | 343 | 81 | |
| GenBank mRNA | BC045404.1 | - | 1 - 2 | 322 | 81 | |
| BC163959.1 | - | 1 - 2 | 140 | 81 | ||
More Information
Set Membership: |
|

amd1
Genomic Map