Assay Details
| Gene Symbol: minpp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201100.1 | NP_957394.1 | 2 - 3 | 846 | 87 | |
| GenBank mRNA | BC047824.1 | - | 2 - 3 | 846 | 87 | |
| BC066753.1 | - | 2 - 3 | 820 | 87 | ||
| BC165733.1 | - | 2 - 3 | 807 | 87 | ||
More Information
Set Membership: |
|

minpp1b
Genomic Map