Assay Details
| Gene Symbol: dtnbp1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201134.1 | NP_957428.1 | 5 - 6 | 416 | 77 | |
| XM_005158097.3 | XP_005158154.1 | 5 - 6 | 683 | 77 | ||
| XM_005158098.4 | XP_005158155.1 | 5 - 6 | 681 | 77 | ||
| XM_005158099.4 | XP_005158156.1 | 5 - 6 | 680 | 77 | ||
| XM_005158100.3 | XP_005158157.1 | 5 - 6 | 684 | 77 | ||
| XM_005158101.3 | XP_005158158.1 | 5 - 6 | 683 | 77 | ||
| XM_021466652.1 | XP_021322327.1 | 5 - 6 | 686 | 77 | ||
| GenBank mRNA | BC049451.1 | - | 5 - 6 | 416 | 77 | |
| BC152672.1 | - | 5 - 6 | 654 | 77 | ||
More Information
Set Membership: |
|

dtnbp1a
Genomic Map