Assay Details
| Gene Symbol: dtnbp1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201134.1 | NP_957428.1 | 8 - 9 | 727 | 107 | |
| XM_005158097.3 | XP_005158154.1 | 8 - 9 | 994 | 107 | ||
| XM_005158098.4 | XP_005158155.1 | 8 - 9 | 992 | 107 | ||
| XM_005158099.4 | XP_005158156.1 | 8 - 9 | 991 | 107 | ||
| XM_005158105.4 | XP_005158162.1 | 2 - 3 | 385 | 107 | ||
| XM_005158106.3 | XP_005158163.1 | 2 - 3 | 416 | 107 | ||
| GenBank mRNA | BC049451.1 | - | 8 - 9 | 727 | 107 | |
| BC152672.1 | - | 8 - 9 | 965 | 107 | ||
More Information
Set Membership: |
|

dtnbp1a
Genomic Map