Assay Details
| Gene Symbol: dtnbp1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201134.1 | NP_957428.1 | 9 - 10 | 872 | 75 | |
| XM_005158099.4 | XP_005158156.1 | 9 - 10 | 1136 | 75 | ||
| XM_005158101.3 | XP_005158158.1 | 9 - 10 | 1130 | 75 | ||
| XM_005158106.3 | XP_005158163.1 | 3 - 4 | 561 | 75 | ||
| XM_005158108.4 | XP_005158165.1 | 3 - 4 | 521 | 75 | ||
| GenBank mRNA | BC049451.1 | - | 9 - 10 | 872 | 75 | |
| BC152672.1 | - | 9 - 10 | 1110 | 75 | ||
More Information
Set Membership: |
|

dtnbp1a
Genomic Map