Assay Details
| Gene Symbol: txnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201138.2 | NP_957432.2 | 1 - 2 | 256 | 62 | |
| XM_021468785.1 | XP_021324460.1 | 1 - 2 | 230 | 62 | ||
| GenBank mRNA | BC045322.1 | - | 1 - 2 | 256 | 62 | |
| BC164656.1 | - | 1 - 2 | 134 | 62 | ||
More Information
Set Membership: |
|

txnl1
Genomic Map