Assay Details
| Gene Symbol: ppih | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201205.1 | NP_957499.1 | 8 - 9 | 564 | 114 | |
| GenBank mRNA | BC045448.1 | - | 8 - 9 | 564 | 114 | |
More Information
Set Membership: |
|

ppih
Genomic Map