Assay Details
| Gene Symbol: grinab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001308611.1 | NP_001295540.1 | 5 - 6 | 868 | 88 | |
| XM_005158451.1 | XP_005158508.1 | 6 - 7 | 1007 | 88 | ||
| GenBank mRNA | BC053253.1 | - | 5 - 6 | 773 | 88 | |
| BC152278.1 | - | 5 - 6 | 782 | 88 | ||
| EH562651.1 | - | 2 - 3 | 464 | 88 | ||
More Information
Set Membership: |
|

grinab
Genomic Map