Assay Details
| Gene Symbol: ube2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201273.1 | NP_958430.1 | 4 - 5 | 367 | 87 | |
| XM_009291145.3 | XP_009289420.1 | 4 - 5 | 438 | 87 | ||
| GenBank mRNA | BC053256.1 | - | 4 - 5 | 367 | 87 | |
More Information
Set Membership: |
|

ube2a
Genomic Map