Assay Details
| Gene Symbol: pgam1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201300.2 | NP_958457.2 | 1 - 2 | 274 | 75 | |
| GenBank mRNA | BC054936.1 | - | 1 - 2 | 75 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pgam1b| Gene Symbol: pgam1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201300.2 | NP_958457.2 | 1 - 2 | 274 | 75 | |
| GenBank mRNA | BC054936.1 | - | 1 - 2 | 75 | ||
Set Membership: |
|