Assay Details
| Gene Symbol: pgam1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201300.2 | NP_958457.2 | 3 - 4 | 724 | 70 | |
| GenBank mRNA | BC054936.1 | - | 3 - 4 | 724 | 70 | |
More Information
Set Membership: |
|

pgam1b
Genomic Map