Assay Details
| Gene Symbol: sec22bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328526.1 | NP_001315455.1 | 4 - 5 | 500 | 77 | |
| NM_201302.2 | NP_958459.1 | 3 - 4 | 488 | 77 | ||
| GenBank mRNA | BC055515.1 | - | 3 - 4 | 444 | 77 | |
| BC152220.1 | - | 4 - 5 | 487 | 77 | ||
| EH489983.1 | - | 3 - 4 | 477 | 77 | ||
| EH528230.1 | - | 2 - 3 | 551 | 77 | ||
More Information
Set Membership: |
|

sec22bb
Genomic Map