Assay Details
| Gene Symbol: pmp22a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201311.1 | NP_958468.1 | 5 - 6 | 652 | 75 | |
| XM_005164439.4 | XP_005164496.1 | 4 - 5 | 402 | 75 | ||
| XM_005164440.4 | XP_005164497.1 | 4 - 5 | 493 | 75 | ||
| GenBank mRNA | BC062526.1 | - | 5 - 6 | 652 | 75 | |
| BC153614.1 | - | 4 - 5 | 456 | 75 | ||
More Information
Set Membership: |
|

pmp22a
Genomic Map