Assay Details
| Gene Symbol: gfi1ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201338.1 | NP_958495.1 | 5 - 6 | 1007 | 117 | |
| GenBank mRNA | AY302246.1 | - | 5 - 6 | 1007 | 117 | |
| BC163682.1 | - | 5 - 6 | 1048 | 117 | ||
More Information
Set Membership: |
|

gfi1ab
Genomic Map