Assay Details
Gene Symbol: mmp13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF506756.1 | - | 4 - 5 | 650 | 91 | |
BC049472.1 | - | 4 - 5 | 598 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
