Assay Details
Gene Symbol: mmp13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF506756.1 | - | 6 - 7 | 940 | 67 | |
BC049472.1 | - | 6 - 7 | 888 | 67 | ||
BC065591.1 | - | 6 - 7 | 938 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
