Assay Details
Gene Symbol: bicc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203420.1 | NP_981965.1 | 15 - 16 | 2078 | 107 | |
GenBank mRNA | AY513238.1 | - | 15 - 16 | 2078 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
