Assay Details
| Gene Symbol: postnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203421.1 | NP_981966.1 | 18 - 19 | 2266 | 103 | |
| XM_021481350.1 | XP_021337025.1 | 19 - 20 | 2473 | 103 | ||
| GenBank mRNA | AB126325.1 | - | 18 - 19 | 2266 | 103 | |
| BC162274.1 | - | 18 - 19 | 2320 | 103 | ||
More Information
Set Membership: |
|

postnb
Genomic Map