Assay Details
| Gene Symbol: postnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203421.1 | NP_981966.1 | 19 - 20 | 2302 | 75 | |
| XM_021481350.1 | XP_021337025.1 | 20 - 21 | 2509 | 75 | ||
| GenBank mRNA | AB126325.1 | - | 19 - 20 | 2302 | 75 | |
| BC162274.1 | - | 19 - 20 | 2356 | 75 | ||
More Information
Set Membership: | 
		   
					
						
							 | 
				

postnb
Genomic Map